Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152594.2 | NP_689807.1 | 7 | 1720 | |
XM_005254202.3 | XP_005254259.1 | 8 | 2520 | ||
XM_011521289.2 | XP_011519591.1 | 7 | 1417 | ||
GenBank mRNA | AK091222.1 | - | 8 | 1490 | |
AK095219.1 | - | 1 | 314 | ||
BC137480.1 | - | 7 | 2245 | 9021480 | |
BC137481.1 | - | 7 | 2245 | 9021481 | |
BX538234.1 | - | 1 | 85 |