Gene Transcripts

Gene Symbol
KRTAP11-1
Entrez Gene ID
337880
Gene Name
keratin associated protein 11-1
Gene Aliases
HACL-1, HACL1, KAP11.1
Chromosome Location
Chr. 21: 30880644 - 30881555 on Build GRCh38
UniGene ID
Hs.407654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175858.2 NP_787054.1 1 542
GenBank mRNA AJ457065.1 - 1 542
BC130555.1 - 1 535 40146586
BC130557.1 - 1 535 40146588

View Full Product Details