Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183387.2 | NP_899243.1 | 5 | 822 | |
XM_006720070.3 | XP_006720133.1 | 5 | 622 | ||
XM_011536528.2 | XP_011534830.1 | 5 | 623 | ||
XM_011536530.2 | XP_011534832.1 | 5 | 622 | ||
XM_011536531.2 | XP_011534833.1 | 5 | 622 | ||
XM_011536532.2 | XP_011534834.1 | 5 | 622 | ||
XM_011536533.2 | XP_011534835.1 | 5 | 622 | ||
XM_011536534.2 | XP_011534836.1 | 6 | 540 | ||
XM_011536536.2 | XP_011534838.1 | 5 | 622 | ||
XM_011536538.2 | XP_011534840.1 | 5 | 622 | ||
XM_017021062.1 | XP_016876551.1 | 5 | 622 | ||
XM_017021063.1 | XP_016876552.1 | 5 | 622 | ||
XM_017021064.1 | XP_016876553.1 | 5 | 622 | ||
XM_017021065.1 | XP_016876554.1 | 5 | 622 | ||
XM_017021066.1 | XP_016876555.1 | 5 | 591 | ||
XM_017021067.1 | XP_016876556.1 | 5 | 943 | ||
XM_017021068.1 | XP_016876557.1 | 5 | 622 | ||
XM_017021069.1 | XP_016876558.1 | 5 | 867 | ||
XM_017021070.1 | XP_016876559.1 | 5 | 622 | ||
GenBank mRNA | BC032685.1 | - | 6 | 770 | |
BC150639.1 | - | 5 | 822 | 9057009 |