Gene Transcripts

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
Chromosome Location
Chr. 10: 59245933 - 59363186 on Build GRCh38
UniGene ID
Hs.607594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001971.2 NP_001001971.1 8 1033
NM_001143773.1 NP_001137245.1 9 1204
NM_001166698.1 NP_001160170.1 9 1118
NM_198215.3 NP_937858.2 8 1033
XM_005269618.4 XP_005269675.2 8 1033
XM_005269619.4 XP_005269676.2 8 1033
XM_006717702.3 XP_006717765.1 10 1034
XM_006717703.3 XP_006717766.1 10 1216
XM_011539490.2 XP_011537792.1 9 1089
XM_011539491.2 XP_011537793.1 10 1135
XM_017015885.1 XP_016871374.1 9 947
XM_017015886.1 XP_016871375.1 9 1204
XM_017015887.1 XP_016871376.1 9 954
XM_017015888.1 XP_016871377.1 10 1216
XM_017015889.1 XP_016871378.1 10 1034
XM_017015890.1 XP_016871379.1 10 1273
XM_017015891.1 XP_016871380.1 9 1204
XM_017015892.1 XP_016871381.1 9 1204
XM_017015893.1 XP_016871382.1 11 1305
GenBank mRNA AK289796.1 - 8 959
AK294805.1 - 8 948
AK299924.1 - 9 1204
AK316542.1 - 9 1114
BC036453.1 - 8 991 5270033
BC064431.1 - 8 1033 5764753
U79304.1 - 6 641

View Full Product Details