Gene Transcripts

Gene Symbol
MOAP1
Entrez Gene ID
64112
Gene Name
modulator of apoptosis 1
Gene Aliases
MAP-1, PNMA4
Chromosome Location
Chr. 14: 93182196 - 93184904 on Build GRCh38
UniGene ID
Hs.24719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022151.4 NP_071434.2 3 989
GenBank mRNA AF305550.1 - 1 619
AK024029.1 - 3 961
AK315522.1 - 3 1001
BC015044.1 - 3 923 3922055

View Full Product Details