Gene Transcripts

Gene Symbol
C1orf21
Entrez Gene ID
81563
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
PIG13
Chromosome Location
Chr. 1: 184387016 - 184629021 on Build GRCh38
UniGene ID
Hs.497159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030806.3 NP_110433.1 6 925
GenBank mRNA AF312864.1 - 6 847
AK056487.1 - 6 661
AY258286.1 - 6 838
BC028567.2 - 6 883 3635521

View Full Product Details