Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001131028.1 | NP_001124500.1 | 6 | 867 | |
NM_031482.4 | NP_113670.1 | 5 | 732 | ||
XM_005248610.4 | XP_005248667.1 | 5 | 2002 | ||
XM_005248611.4 | XP_005248668.1 | 5 | 576 | ||
XM_005248612.3 | XP_005248669.1 | 4 | 628 | ||
XM_011543660.2 | XP_011541962.1 | 4 | 432 | ||
XM_011543661.2 | XP_011541963.1 | 3 | 815 | ||
XM_017009944.1 | XP_016865433.1 | 5 | 763 | ||
GenBank mRNA | AK225969.1 | - | 4 | 624 | |
AK315752.1 | - | 5 | 732 | ||
AL136912.1 | - | 5 | 600 | ||
BC029268.1 | - | 5 | 702 | 4521833 |