Gene Transcripts

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location
Chr. 11: 19345200 - 20121601 on Build GRCh38
UniGene ID
Hs.502116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244963.1 NP_001231892.1 1 512
NM_145117.4 NP_660093.2 1 512
NM_182964.5 NP_892009.3 1 512
XM_006718364.3 XP_006718427.1 1 834
XM_006718365.3 XP_006718428.1 1 834
XM_011520444.2 XP_011518746.1 1 834
XM_011520445.2 XP_011518747.1 1 834
XM_011520446.2 XP_011518748.1 1 834
XM_011520447.2 XP_011518749.1 1 834
XM_011520449.2 XP_011518751.1 1 834
XM_011520450.2 XP_011518752.1 1 834
XM_017018521.1 XP_016874010.1 1 834
GenBank mRNA AB037840.3 - 1 513
AB063116.1 - 1 481
AF466143.1 - 1 227
AF466144.1 - 1 227
AJ488102.1 - 1 250
BC150290.1 - 1 513 8860130

View Full Product Details