Gene Transcripts

Gene Symbol
EPB41L4B
Entrez Gene ID
54566
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases
CG1, EHM2
Chromosome Location
Chr. 9: 109171974 - 109320964 on Build GRCh38
UniGene ID
Hs.591901
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018424.3 NP_060894.2 4 986
NM_019114.4 NP_061987.3 4 986
XM_011518790.1 XP_011517092.1 4 483
XM_011518791.2 XP_011517093.1 4 468
XM_011518792.2 XP_011517094.1 4 468
XM_011518794.2 XP_011517096.1 4 468
XM_017014813.1 XP_016870302.1 4 468
XM_017014814.1 XP_016870303.1 4 468
XM_017014815.1 XP_016870304.1 4 468
GenBank mRNA AB032179.3 - 4 986
AF153416.1 - 4 758
AF153418.1 - 4 758
AK000388.1 - 4 370

View Full Product Details