Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
Chromosome Location
Chr. 1: 35720465 - 35769985 on Build GRCh38
UniGene ID
Hs.175613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190481.1 NP_001177410.1 5 874
NM_022111.3 NP_071394.2 5 874
XM_011541938.2 XP_011540240.1 5 897
XM_011541939.2 XP_011540241.1 5 891
XM_011541940.2 XP_011540242.1 5 891
XM_011541941.2 XP_011540243.1 4 729
GenBank mRNA AF297866.1 - 5 874
AK308193.1 - 5 903
BC018670.1 - 5 894
BC038991.2 - 5 900
BC113116.1 - 5 867 7961377
BC115025.1 - 5 867 7961363
BC115026.1 - 5 867 7961366
BC137279.1 - 5 853 9021276
BC140789.1 - 5 853 9021657

View Full Product Details