Gene Transcripts

Gene Symbol
ITGB1BP2
Entrez Gene ID
26548
Gene Name
integrin subunit beta 1 binding protein 2
Gene Aliases
CHORDC3, ITGB1BP, MELUSIN, MSTP015
Chromosome Location
Chr. X: 71301275 - 71305371 on Build GRCh38
UniGene ID
Hs.109999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012278.2 NP_036410.1 3 202
GenBank mRNA AF110225.1 - 3 156
AF140690.1 - 3 172
BC108901.2 - 2 429 40004692
DA895605.1 - 3 205

View Full Product Details