Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304476.1 | NP_001291405.1 | 3 | 694 | |
NM_001304477.1 | NP_001291406.1 | 2 | 199 | ||
NM_014178.7 | NP_054897.4 | 2 | 841 | ||
XM_017021231.1 | XP_016876720.1 | 2 | 573 | ||
XM_017021232.1 | XP_016876721.1 | 3 | 3169 | ||
XM_017021233.1 | XP_016876722.1 | 3 | 521 | ||
XM_017021234.1 | XP_016876723.1 | 3 | 294 | ||
XM_017021235.1 | XP_016876724.1 | 2 | 3082 | ||
XM_017021236.1 | XP_016876725.1 | 3 | 498 | ||
XM_017021237.1 | XP_016876726.1 | 3 | 327 | ||
XM_017021238.1 | XP_016876727.1 | 2 | 265 | ||
XM_017021239.1 | XP_016876728.1 | 2 | 263 | ||
XM_017021240.1 | XP_016876729.1 | 2 | 185 | ||
XM_017021241.1 | XP_016876730.1 | 2 | 802 | ||
XM_017021242.1 | XP_016876731.1 | 3 | 267 | ||
GenBank mRNA | AF391153.1 | - | 1 | 165 | |
AK096957.1 | - | 3 | 262 | ||
AK309203.1 | - | 2 | 199 | ||
AL834346.1 | - | 2 | 179 | ||
BC009499.2 | - | 3 | 468 | 3955370 | |
BC067278.1 | - | 2 | 841 | 6178786 | |
BM738712.1 | - | 2 | 429 |