Gene Transcripts

Gene Symbol
ZNF330
Entrez Gene ID
27309
Gene Name
zinc finger protein 330
Gene Aliases
HSA6591, NOA36
Chromosome Location
Chr. 4: 141220294 - 141234697 on Build GRCh38
UniGene ID
Hs.120766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292002.1 NP_001278931.1 8, 9 840
NM_014487.5 NP_055302.1 9, 10 911
XM_017008033.1 XP_016863522.1 9, 10 1055
GenBank mRNA AJ006591.2 - 9, 10 856
AK002106.1 - 9, 10 860
AK223609.1 - 9, 10 862
AK294089.1 - 8, 9 794
AK315463.1 - 9, 10 897
BC004421.1 - 9, 10 887 3352916

View Full Product Details