Gene Transcripts

Gene Symbol
P2RY10
Entrez Gene ID
27334
Gene Name
purinergic receptor P2Y10
Gene Aliases
LYPSR2, P2Y10
Chromosome Location
Chr. X: 78945332 - 78963727 on Build GRCh38
UniGene ID
Hs.296433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324218.1 NP_001311147.1 3 444
NM_001324221.1 NP_001311150.1 3 350
NM_001324225.1 NP_001311154.1 4 429
NM_014499.3 NP_055314.1 4 493
NM_198333.2 NP_938147.1 2 301
GenBank mRNA AK292138.1 - 4 431
AK313218.1 - 2 242
BC051875.1 - 4 404 5212240
BC095498.1 - 1 129 6971933
CD639881.1 - 2 282
DA817144.1 - 4 370
DB131258.1 - 3 347

View Full Product Details