Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015474.3 | NP_056289.2 | 5 | 734 | |
XM_005260384.3 | XP_005260441.1 | 5 | 707 | ||
XM_011528761.2 | XP_011527063.1 | 5 | 707 | ||
GenBank mRNA | AB013847.1 | - | 5 | 534 | |
AB208944.1 | - | 5 | 564 | ||
AF228421.1 | - | 5 | 687 | ||
AK027811.1 | - | 5 | 707 | ||
AK304187.1 | - | 5 | 491 | ||
AK304795.1 | - | 6 | 662 | ||
AK315169.1 | - | 5 | 667 | ||
AL050267.1 | - | 5 | 609 | ||
BC036450.2 | - | 5 | 562 | 4816438 |