Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142541.2 | NP_001136013.1 | 6 | 800 | |
NM_015948.4 | NP_057032.2 | 6 | 1162 | ||
NR_109913.1 | 6 | 1011 | |||
NR_109914.1 | 4 | 1096 | |||
XM_005249156.3 | XP_005249213.1 | 5 | 942 | ||
XM_005249157.2 | XP_005249214.1 | 6 | 935 | ||
XM_005249158.1 | XP_005249215.1 | 5 | 830 | ||
XM_006715102.3 | XP_006715165.1 | 6 | 972 | ||
XM_006715103.3 | XP_006715166.1 | 5 | 942 | ||
XM_011514648.1 | XP_011512950.1 | 7 | 942 | ||
XM_011514650.2 | XP_011512952.1 | 5 | 942 | ||
XM_017010910.1 | XP_016866399.1 | 6 | 1258 | ||
XM_017010911.1 | XP_016866400.1 | 5 | 1176 | ||
XM_017010912.1 | XP_016866401.1 | 6 | 890 | ||
XM_017010913.1 | XP_016866402.1 | 6 | 1258 | ||
XM_017010914.1 | XP_016866403.1 | 5 | 1026 | ||
XM_017010915.1 | XP_016866404.1 | 5 | 830 | ||
GenBank mRNA | AB231931.1 | - | 4 | 637 | |
AF132953.1 | - | 5 | 658 | ||
AK313522.1 | - | 6 | 778 | ||
BC006973.1 | - | 6 | 988 | 3827889 | |
BX538271.1 | - | 6 | 1006 | ||
BX641086.1 | - | 4 | 6159 | ||
BX647208.1 | - | 4 | 1030 | ||
BX648022.1 | - | 5 | 5099 |