Gene Transcripts

Gene Symbol
DBR1
Entrez Gene ID
51163
Gene Name
debranching RNA lariats 1
Gene Aliases
Chromosome Location
Chr. 3: 138160988 - 138174949 on Build GRCh38
UniGene ID
Hs.477700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016216.3 NP_057300.2 5 657
GenBank mRNA AF180919.2 - 5 611
AK094147.1 - 6 542
AL702928.1 - 5 659
BC009472.1 - 5 585 3503627

View Full Product Details