Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277174.1 | NP_001264103.1 | 6 | 2439 | |
NM_005864.3 | NP_005855.1 | 6 | 2946 | ||
NM_032459.2 | NP_115835.1 | 5 | 2667 | ||
XM_005267256.1 | XP_005267313.1 | 5 | 2437 | ||
GenBank mRNA | AB001466.1 | - | 6 | 2946 |