Gene Transcripts

Gene Symbol
STYK1
Entrez Gene ID
55359
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
NOK, SuRTK106
Chromosome Location
Chr. 12: 10618923 - 10674318 on Build GRCh38
UniGene ID
Hs.24979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018423.2 NP_060893.2 11 2654
XM_005253417.2 XP_005253474.1 10 2554
XM_011520736.1 XP_011519038.1 12 2457
XM_011520737.2 XP_011519039.1 11 2494
XM_011520738.2 XP_011519040.1 11 2649
XM_017019579.1 XP_016875068.1 11 2437
GenBank mRNA AF251059.1 - 11 2652
AL353940.1 - 11 2402

View Full Product Details