Gene Transcripts
- Gene Symbol
- C6orf10
- Entrez Gene ID
- 10665
- Gene Name
- chromosome 6 open reading frame 10
- Gene Aliases
- TSBP
- Chromosome Location
- Chr. 6: 32290523 - 32371912 on Build GRCh38
- UniGene ID
- Hs.567414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286474.1 |
NP_001273403.1
|
12, 13 |
566 |
|
NM_001286475.1 |
NP_001273404.1
|
11, 12 |
545 |
|
NM_006781.4 |
NP_006772.3
|
11, 12 |
614 |
|
XM_017010182.1 |
XP_016865671.1
|
|
12 |
|
XM_017010183.1 |
XP_016865672.1
|
1, 2 |
575 |
|
XM_017010184.1 |
XP_016865673.1
|
1, 2 |
575 |
|
XM_017010185.1 |
XP_016865674.1
|
12, 13 |
639 |
|
GenBank mRNA |
AK225742.1 |
- |
12, 13 |
557 |
|
AK302176.1 |
- |
11, 12 |
505 |
|
AK310230.1 |
- |
11, 12 |
534 |
|
BC034774.1 |
- |
11, 12 |
581 |
5169157 |
U60665.1 |
- |
12, 13 |
596 |
|
View Full Product Details