Gene Transcripts

Gene Symbol
BTG3
Entrez Gene ID
10950
Gene Name
BTG anti-proliferation factor 3
Gene Aliases
ANA, APRO4, TOB5, TOB55, TOFA
Chromosome Location
Chr. 21: 17593650 - 17612950 on Build GRCh38
UniGene ID
Hs.473420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130914.1 NP_001124386.1 2, 3 428
NM_006806.4 NP_006797.3 2, 3 428
XM_011529441.1 XP_011527743.1 3, 4 536
XM_017028265.1 XP_016883754.1 3, 4 566
XM_017028266.1 XP_016883755.1 3, 4 523
GenBank mRNA AK289408.1 - 2, 3 376
BC011957.1 - 2, 3 322 3859781
BT007276.1 - 1, 2 168
CR457061.1 - 1, 2 168
D64110.1 - 2, 3 262
DA381239.1 - 2, 3 429
DR155504.1 - 2, 3 344

View Full Product Details