Gene Transcripts

Gene Symbol
PIM2
Entrez Gene ID
11040
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
Chromosome Location
Chr. X: 48913182 - 48919136 on Build GRCh38
UniGene ID
Hs.727148
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006875.3 NP_006866.2 6 2049
GenBank mRNA BC018111.1 - 6 1916
U77735.1 - 6 1933

View Full Product Details