Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320800.1 | NP_001307729.1 | 10 | 2338 | |
NM_007170.2 | NP_009101.2 | 11 | 2646 | ||
XM_005270355.3 | XP_005270412.1 | 10 | 2471 | ||
XM_006710287.2 | XP_006710350.1 | 12 | 2686 | ||
XM_011540497.1 | XP_011538799.1 | 13 | 2719 | ||
XM_017000047.1 | XP_016855536.1 | 11 | 2704 | ||
XM_017000048.1 | XP_016855537.1 | 12 | 2974 | ||
XM_017000049.1 | XP_016855538.1 | 10 | 2771 | ||
GenBank mRNA | AB057597.1 | - | 11 | 2636 | |
AJ132545.1 | - | 10 | 2588 | ||
AK027573.1 | - | 11 | 2620 | ||
AK294172.1 | - | 10 | 2313 | ||
BC033085.1 | - | 9 | 2170 | 5425494 | |
BX648441.1 | - | 5 | 1804 |