Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145047.1 | NP_001138519.1 | 3 | 180 | |
NM_001145048.1 | NP_001138520.1 | 2 | 112 | ||
NM_001145049.1 | NP_001138521.1 | 2 | 112 | ||
NM_007266.3 | NP_009197.2 | 2 | 219 | ||
NR_026735.1 | 2 | 219 | |||
GenBank mRNA | AB044661.1 | - | 2 | 180 | |
AJ010842.1 | - | 2 | 109 | ||
AK298827.1 | - | 2 | 513 | ||
AK298866.1 | - | 2 | 219 | ||
AK302131.1 | - | 2 | 185 | ||
AK310340.1 | - | 3 | 170 | ||
AK311423.1 | - | 2 | 242 | ||
AK312992.1 | - | 2 | 191 | ||
AK316404.1 | - | 2 | 112 | ||
BC007451.2 | - | 2 | 163 | 3029376 |