Gene Transcripts

Gene Symbol
CCNDBP1
Entrez Gene ID
23582
Gene Name
cyclin D1 binding protein 1
Gene Aliases
DIP1, GCIP, HHM
Chromosome Location
Chr. 15: 43185118 - 43197177 on Build GRCh38
UniGene ID
Hs.36794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012142.4 NP_036274.3 8 944
NR_027513.2 6 767
NR_027514.2 7 847
NR_045998.1 7 864
NR_045999.1 5 707
XM_006720448.1 XP_006720511.1 6 866
GenBank mRNA AF082569.1 - 8 803
AF087852.1 - 8 789
AF113535.1 - 6 753
AF132034.1 - 8 759
AF246144.1 - 8 761
AK075146.1 - 8 773
AK075296.1 - 6 758
AK124193.1 - 3 1348
AK128849.1 - 4 1988
AK225143.1 - 8 759
AK225645.1 - 8 773
AK290665.1 - 8 773
AK295017.1 - 7 676
AL552334.3 - 8 882
BC009689.1 - 8 787 3896421
BQ439439.1 - 5 521
CR450331.1 - 8 713

View Full Product Details