Gene Transcripts

Gene Symbol
FBXW10
Entrez Gene ID
10517
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
Chromosome Location
Chr. 17: 18744013 - 18779349 on Build GRCh38
UniGene ID
Hs.592128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267585.1 NP_001254514.1 5 1255
NM_001267586.1 NP_001254515.1 5 1255
NR_051988.1 5 1901
XM_011523625.1 XP_011521927.1 4 1408
XM_011523626.2 XP_011521928.1 4 1408
XM_017024020.1 XP_016879509.1 4 1408
XM_017024021.1 XP_016879510.1 4 1408
GenBank mRNA AK310383.1 - 5 1840
AL136822.1 - 5 1193
AY729024.1 - 5 1023
BC028364.2 - 4 1329 4827344

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
Chromosome Location
Chr. 17: 15565482 - 15619704 on Build GRCh38
UniGene ID
Hs.548021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282540.1 NP_001269469.1 5 1215
NM_006382.3 NP_006373.2 5 1215
GenBank mRNA CB989113.1 - 3 519

View Full Product Details