Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004884.3 | NP_004875.2 | 12, 13 | 2257 | |
XM_011522241.1 | XP_011520543.1 | 12, 13 | 2199 | ||
XM_011522243.1 | XP_011520545.1 | 11, 12 | 1783 | ||
XM_011522244.1 | XP_011520546.1 | 11, 12 | 1772 | ||
GenBank mRNA | AF063936.1 | - | 4, 5 | 489 | |
AK095529.1 | - | 11, 12 | 1772 | ||
BC042054.1 | - | 12, 13 | 2205 | 5579570 | |
BC067107.1 | - | 12, 13 | 2257 | 6644528 | |
BX389424.1 | - | 2, 3 | 194 |