Gene Transcripts

Gene Symbol
ZMYM4
Entrez Gene ID
9202
Gene Name
zinc finger MYM-type containing 4
Gene Aliases
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location
Chr. 1: 35268694 - 35421944 on Build GRCh38
UniGene ID
Hs.269211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005095.2 NP_005086.2 5 886
XM_005271328.4 XP_005271385.1 5 1165
XM_011542421.2 XP_011540723.1 5 812
XM_011542423.2 XP_011540725.1 6 1217
XM_017002803.1 XP_016858292.1 6 939
XM_017002804.1 XP_016858293.1 5 803
XM_017002805.1 XP_016858294.1 3 718
GenBank mRNA AK308320.1 - 5 886
AK309705.1 - 4 546
BC012093.1 - 3 465
BC127113.1 - 6 892 40125110
BX647376.1 - 1 116
CR936741.1 - 4 750

View Full Product Details