Gene Transcripts

Gene Symbol
RBM8A
Entrez Gene ID
9939
Gene Name
RNA binding motif protein 8A
Gene Aliases
BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
Chromosome Location
Chr. 1: 145921556 - 145927536 on Build GRCh38
UniGene ID
Hs.591455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005105.4 NP_005096.1 6 2795
GenBank mRNA AF127761.1 - 6 2697
AK075009.1 - 6 2716
AL049219.1 - 1 180
BC071577.1 - 6 2742

View Full Product Details