Gene Transcripts

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
Chromosome Location
Chr. 14: 24171853 - 24180408 on Build GRCh38
UniGene ID
Hs.419259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048205.1 NP_001041670.1 19 2120
NM_005132.2 NP_005123.2 20 2019
XM_011537421.1 XP_011535723.1 18 2395
XM_017021841.1 XP_016877330.1 18 2269
GenBank mRNA AF006264.1 - 19 1592
AK001435.1 - 20 2019
AK291191.1 - 20 2151
AL832973.1 - 17 2509
BC004159.2 - 19 1676 2822680
BC010887.1 - 19 2053 3535425
BC019326.1 - 13 1038
CR457263.1 - 19 1592

View Full Product Details