Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr. 12: 49322135 - 49331733 on Build GRCh38
UniGene ID
Hs.524399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005480.3 NP_005471.3 9, 10 1197
XM_006719181.1 XP_006719244.1 9, 10 1141
XM_011537723.1 XP_011536025.1 9, 10 1141
XM_011537724.1 XP_011536026.1 8, 9 937
GenBank mRNA AK299302.1 - 9, 10 1004
AK309716.1 - 6, 7 618
AK310709.1 - 8, 9 935
AK315211.1 - 9, 10 1141
BC032583.1 - 9, 10 1109 5498800
U04810.2 - 9, 10 1140

View Full Product Details