Gene Transcripts

Gene Symbol
STK11
Entrez Gene ID
6794
Gene Name
serine/threonine kinase 11
Gene Aliases
LKB1, PJS, hLKB1
Chromosome Location
Chr. 19: 1205799 - 1228435 on Build GRCh38
UniGene ID
Hs.515005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000455.4 NP_000446.1 2 1431
XM_005259617.2 XP_005259674.1 2 941
XM_005259618.3 XP_005259675.1 2 941
XM_011528209.1 XP_011526511.1 3 1072
GenBank mRNA AB209553.1 - 2 406
AF035625.1 - 2 654
AF217978.1 - 3 1277
AK314858.1 - 2 442
BC007981.2 - 2 766 3689780
BC019334.1 - 2 587 3946005
EF644172.1 - 2 400
GU573916.1 - 1 24
KU178339.1 - 2 316
U63333.1 - 2 316

View Full Product Details