Gene Transcripts

Gene Symbol
ZAN
Entrez Gene ID
7455
Gene Name
zonadhesin (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 7: 100733599 - 100797797 on Build GRCh38
UniGene ID
Hs.307004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003386.2 NP_003377.2 34 6424
NM_173059.2 NP_775082.2 34 6424
NR_111917.1 34 6424
NR_111918.1 34 6424
NR_111919.1 34 6424
XM_011516555.2 XP_011514857.1 34 6157
XM_017012581.1 XP_016868070.1 21 3913
GenBank mRNA AF332975.1 - 34 6424
AF332976.1 - 34 6424
AF332977.1 - 34 6424
AF332978.1 - 34 6424
AF332979.1 - 34 6424
AF332980.1 - 34 6424
AK302596.1 - 13 1854

View Full Product Details