Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197323.1 | NP_001184252.1 | 6 | 1251 | |
NM_003609.4 | NP_003600.2 | 7 | 2189 | ||
GenBank mRNA | AJ223349.1 | - | 6 | 745 | |
AJ223350.1 | - | 6 | 1567 | ||
AJ223351.2 | - | 7 | 2086 | ||
AK075145.1 | - | 6 | 1502 | ||
BC000588.2 | - | 7 | 2159 | 3345739 |