Gene Transcripts

Gene Symbol
ZFAND5
Entrez Gene ID
7763
Gene Name
zinc finger AN1-type containing 5
Gene Aliases
ZA20D2, ZFAND5A, ZNF216
Chromosome Location
Chr. 9: 72351425 - 72365247 on Build GRCh38
UniGene ID
Hs.406096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102420.2 NP_001095890.1 4 911
NM_001102421.2 NP_001095891.1 3 774
NM_001278243.1 NP_001265172.1 4 490
NM_001278244.1 NP_001265173.1 3 353
NM_001278245.1 NP_001265174.1 4 487
NM_006007.3 NP_005998.1 3 771
GenBank mRNA AF062346.1 - 3 494
AF062347.1 - 3 349
AK025782.1 - 4 374
AK290849.1 - 3 494
AK307376.1 - 1 195
BC011018.1 - 3 610 4331933
BC027707.2 - 4 861 3945047
BC073131.1 - 3 275 6137219
DA290666.1 - 3 353
DA392231.1 - 4 418
DA914616.1 - 4 434

View Full Product Details