Gene Transcripts

Gene Symbol
FBP2
Entrez Gene ID
8789
Gene Name
fructose-bisphosphatase 2
Gene Aliases
Chromosome Location
Chr. 9: 94558714 - 94593832 on Build GRCh38
UniGene ID
Hs.61255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003837.3 NP_003828.2 7 1233
GenBank mRNA AJ573829.1 - 1 305
Y10812.2 - 7 1194

View Full Product Details