Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286673.1 | NP_001273602.1 | 5 | 1129 | |
NM_001286674.1 | NP_001273603.1 | 3 | 896 | ||
NM_001286675.1 | NP_001273604.1 | 4 | 980 | ||
NM_003702.4 | NP_003693.2 | 5 | 1085 | ||
NM_170587.3 | NP_733466.1 | 6 | 1474 | ||
NR_104579.1 | 4 | 936 | |||
XM_011517622.2 | XP_011515924.1 | 5 | 1393 | ||
GenBank mRNA | AF060877.2 | - | 5 | 951 | |
AF074979.1 | - | 4 | 869 | ||
AF366054.1 | - | 5 | 1036 | ||
AF366055.1 | - | 4 | 887 | ||
AF366056.1 | - | 3 | 803 | ||
AF366057.1 | - | 5 | 901 | ||
AK094503.1 | - | 4 | 936 | ||
AY046538.1 | - | 6 | 1473 | ||
BC015614.2 | - | 5 | 922 | ||
BC063490.1 | - | 5 | 1075 | 5240001 |