Gene Transcripts

Gene Symbol
TMEM87A
Entrez Gene ID
25963
Gene Name
transmembrane protein 87A
Gene Aliases
Chromosome Location
Chr. 15: 42210452 - 42273584 on Build GRCh38
UniGene ID
Hs.511138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286487.1 NP_001273416.1 12 1294
NM_015497.4 NP_056312.2 13 1355
XM_005254287.1 XP_005254344.1 13 1209
XM_006720468.2 XP_006720531.1 13 1214
XM_011521443.2 XP_011519745.1 12 1126
XM_017022067.1 XP_016877556.1 13 1211
GenBank mRNA AF132733.1 - 13 1211
AK074870.1 - 13 1204
AK075403.1 - 13 1241
AK289591.1 - 13 1204
AK290150.1 - 13 1190
AK290484.1 - 12 1138
AK297816.1 - 13 1200
AK309915.1 - 13 1204

View Full Product Details