Gene Transcripts

Gene Symbol
KYAT1
Entrez Gene ID
883
Gene Name
kynurenine aminotransferase 1
Gene Aliases
CCBL1, GTK, KAT1, KATI
Chromosome Location
Chr. 9: 128832942 - 128882551 on Build GRCh38
UniGene ID
Hs.495250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122671.1 NP_001116143.1 4 462
NM_001287390.1 NP_001274319.1 6 859
NM_004059.4 NP_004050.3 4 468
NR_109829.1 4 468
XM_006717311.3 XP_006717374.1 5 654
XM_011519165.2 XP_011517467.1 6 830
XM_011519167.2 XP_011517469.2 4 434
XM_011519168.1 XP_011517470.1 3 293
XM_011519169.1 XP_011517471.1 4 562
XM_011519170.2 XP_011517472.1 4 930
XM_017015260.1 XP_016870749.1 5 614
XM_017015261.1 XP_016870750.1 6 836
XM_017015262.1 XP_016870751.1 6 1325
XM_017015263.1 XP_016870752.1 6 740
XM_017015264.1 XP_016870753.1 5 647
XM_017015265.1 XP_016870754.1 4 1024
XM_017015266.1 XP_016870755.1 5 655
XM_017015267.1 XP_016870756.1 4 476
XM_017015268.1 XP_016870757.1 5 638
GenBank mRNA AK291178.1 - 4 456
AK297995.1 - 6 715
AK314427.1 - 4 338
AU123740.1 - 4 446
BC021262.2 - 4 429 4137942
DA381449.1 - 4 468
X82224.1 - 3 283

View Full Product Details