Gene Transcripts
- Gene Symbol
- SSH2
- Entrez Gene ID
- 85464
- Gene Name
- slingshot protein phosphatase 2
- Gene Aliases
- SSH-2, SSH-2L
- Chromosome Location
- Chr. 17: 29625938 - 29930239 on Build GRCh38
- UniGene ID
- Hs.654754
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282129.1 |
NP_001269058.1
|
4, 5 |
521 |
|
NM_001282130.1 |
NP_001269059.1
|
3, 4 |
440 |
|
NM_001282131.1 |
NP_001269060.1
|
2, 3 |
388 |
|
NM_033389.3 |
NP_203747.2
|
3, 4 |
252 |
|
XM_005258058.3 |
XP_005258115.1
|
5, 6 |
414 |
|
XM_005258059.2 |
XP_005258116.1
|
2, 3 |
388 |
|
XM_006722149.3 |
XP_006722212.1
|
2, 3 |
4019 |
|
XM_011525402.2 |
XP_011523704.1
|
4, 5 |
249 |
|
XM_011525404.2 |
XP_011523706.1
|
2, 3 |
1012 |
|
XM_011525405.2 |
XP_011523707.1
|
2, 3 |
1060 |
|
XM_017025251.1 |
XP_016880740.1
|
4, 5 |
460 |
|
XM_017025252.1 |
XP_016880741.1
|
3, 4 |
440 |
|
XM_017025253.1 |
XP_016880742.1
|
4, 5 |
460 |
|
XM_017025254.1 |
XP_016880743.1
|
4, 5 |
460 |
|
XM_017025255.1 |
XP_016880744.1
|
4, 5 |
460 |
|
GenBank mRNA |
AB072358.1 |
- |
3, 4 |
212 |
|
AB072359.1 |
- |
3, 4 |
423 |
|
AB099290.1 |
- |
3, 4 |
212 |
|
AK296413.1 |
- |
4, 5 |
386 |
|
AK308887.1 |
- |
3, 4 |
319 |
|
BC008941.2 |
- |
3, 4 |
364 |
|
BC011636.2 |
- |
2, 3 |
346 |
|
BC068223.1 |
- |
3, 4 |
364 |
|
DA749059.1 |
- |
3, 4 |
441 |
|
DA861985.1 |
- |
2, 3 |
390 |
|
View Full Product Details