Gene Transcripts

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
Chromosome Location
Chr. 17: 29625938 - 29930239 on Build GRCh38
UniGene ID
Hs.654754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282129.1 NP_001269058.1 16 8997
NM_033389.3 NP_203747.2 15 8728
XM_005258058.3 XP_005258115.1 17 8890
XM_005258060.3 XP_005258117.1 3 7457
XM_006722149.3 XP_006722212.1 14 12495
XM_011525402.2 XP_011523704.1 16 8725
XM_011525404.2 XP_011523706.1 14 9488
XM_011525405.2 XP_011523707.1 14 9536
XM_011525406.2 XP_011523708.1 13 8815
XM_011525407.1 XP_011523709.1 13 8560
XM_017025251.1 XP_016880740.1 16 8933
XM_017025252.1 XP_016880741.1 14 8794
GenBank mRNA AB051512.1 - 3 7546
AB099290.1 - 15 8688
AK094204.1 - 1 2191
AK094429.1 - 1 2792

View Full Product Details