Gene Transcripts

Gene Symbol
MARCH9
Entrez Gene ID
92979
Gene Name
membrane associated ring-CH-type finger 9
Gene Aliases
MARCH-IX, RNF179
Chromosome Location
Chr. 12: 57755098 - 57760410 on Build GRCh38
UniGene ID
Hs.632709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138396.5 NP_612405.2 4 1453
GenBank mRNA AK093897.1 - 2 368
AK313925.1 - 2 1130
BC009489.2 - 2 933 3936171
BC036455.2 - 4 1434 4811410
BC050397.1 - 4 1453

View Full Product Details