Gene Transcripts

Gene Symbol
CCDC43
Entrez Gene ID
124808
Gene Name
coiled-coil domain containing 43
Gene Aliases
Chromosome Location
Chr. 17: 44677437 - 44689797 on Build GRCh38
UniGene ID
Hs.579115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099225.1 NP_001092695.1 1 164
NM_144609.2 NP_653210.2 1 164
GenBank mRNA AK056357.1 - 1 129
AL833590.1 - 1 126
BC023298.1 - 1 125
BC047776.2 - 1 130 6023964
BG706097.1 - 1 173
BX648868.1 - 1 146

View Full Product Details