Gene Transcripts

Gene Symbol
MYO1H
Entrez Gene ID
283446
Gene Name
myosin IH
Gene Aliases
Chromosome Location
Chr. 12: 109388277 - 109448371 on Build GRCh38
UniGene ID
Hs.512008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101421.3 NP_001094891.3 27 2724
XM_011538223.2 XP_011536525.1 28 3232
XM_017019207.1 XP_016874696.1 27 3166
GenBank mRNA AK094906.1 - 4 495
AK298093.1 - 5 572
DC359165.1 - 5 573
DC401393.1 - 1 36

View Full Product Details