Gene Transcripts
- Gene Symbol
- LRRC9
- Entrez Gene ID
- 341883
- Gene Name
- leucine rich repeat containing 9
- Gene Aliases
- Chromosome Location
- Chr. 14: 59919713 - 60063559 on Build GRCh38
- UniGene ID
- Hs.666245
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_075071.1 |
|
3 |
382 |
|
GenBank mRNA |
AK128037.1 |
- |
3 |
382 |
|
View Full Product Details