Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000979.3 | NP_000970.1 | 7 | 831 | |
NM_001270490.1 | NP_001257419.1 | 6 | 744 | ||
NR_073022.1 | 7 | 823 | |||
GenBank mRNA | AK026558.1 | - | 7 | 589 | |
AK091257.1 | - | 1 | 3795 | ||
BC000374.2 | - | 6 | 563 | 2820119 | |
BC009708.2 | - | 7 | 577 | 3878799 | |
BC021743.2 | - | 7 | 589 | 5261251 | |
CD246847.1 | - | 6 | 528 | ||
CD389415.1 | - | 7 | 595 | ||
L11566.1 | - | 7 | 571 |