Gene Transcripts
- Gene Symbol
- SMARCC1
- Entrez Gene ID
- 6599
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
- Gene Aliases
- BAF155, CRACC1, Rsc8, SRG3, SWI3
- Chromosome Location
- Chr. 3: 47585888 - 47781915 on Build GRCh38
- UniGene ID
- Hs.476179
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003074.3 |
NP_003065.3
|
5, 6 |
694 |
|
XM_011534034.1 |
XP_011532336.1
|
5, 6 |
425 |
|
GenBank mRNA |
AK302541.1 |
- |
5, 6 |
469 |
|
BC021862.1 |
- |
5, 6 |
631 |
|
BC032564.1 |
- |
5, 6 |
678 |
|
BC039843.1 |
- |
5, 6 |
598 |
|
BC050564.1 |
- |
5, 6 |
623 |
3912713 |
BC065253.1 |
- |
5, 6 |
663 |
|
BC113465.1 |
- |
5, 6 |
642 |
8322517 |
BC117213.1 |
- |
5, 6 |
642 |
40125764 |
U66615.1 |
- |
5, 6 |
630 |
|
View Full Product Details