Gene Transcripts

Gene Symbol
SKP1
Entrez Gene ID
6500
Gene Name
S-phase kinase-associated protein 1
Gene Aliases
EMC19, OCP-II, OCP2, SKP1A, TCEB1L, p19A
Chromosome Location
Chr. 5: 134156391 - 134177033 on Build GRCh38
UniGene ID
Hs.171626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006930.3 NP_008861.2 2, 3 273
NM_170679.2 NP_733779.1 2, 3 273
GenBank mRNA BC009839.2 - 2, 3 157
BC020798.1 - 2, 3 208 4243711
BC025673.1 - 2, 3 170 5203008
BC065730.1 - 2, 3 194 6672613
BI552966.1 - 2, 3 240
BI670085.1 - 2, 3 278
U33760.1 - 2, 3 187
U37558.1 - 1, 2 55
Z47087.1 - 2, 3 203

Gene Symbol
SKP1P2
Entrez Gene ID
728622
Gene Name
S-phase kinase-associated protein 1 pseudogene 2
Gene Aliases
SKP1B, p19B
Chromosome Location
Chr. 12: 16988747 - 16990628 on Build GRCh38
UniGene ID
Hs.741380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036619.1 1 473
GenBank mRNA AK310444.1 - 1 473

View Full Product Details