Gene Transcripts

Gene Symbol
KRT12
Entrez Gene ID
3859
Gene Name
keratin 12
Gene Aliases
K12
Chromosome Location
Chr. 17: 40861178 - 40867210 on Build GRCh38
UniGene ID
Hs.66739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000223.3 NP_000214.1 1 553
GenBank mRNA AK313747.1 - 1 567
D78367.1 - 1 553

View Full Product Details