Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002143.2 | NP_002134.2 | 4 | 802 | |
XM_005270792.2 | XP_005270849.1 | 4 | 938 | ||
XM_017001118.1 | XP_016856607.1 | 4 | 1274 | ||
GenBank mRNA | BC001777.1 | - | 4 | 802 | |
D16593.1 | - | 3 | 770 |